Assay Details
| Gene Symbol: RAB27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004163.4 | NP_004154.2 | 6 | 2741 | 109 | |
| XM_005266733.1 | XP_005266790.1 | 7 | 2662 | 109 | ||
| XM_017025913.1 | XP_016881402.1 | 9 | 3495 | 109 | ||
| XM_017025914.1 | XP_016881403.1 | 8 | 2890 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586570.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000586594.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007360.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150489.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150489.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150489.hg.1 | TC1800007360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800150489.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150489.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150491.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150491.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150491.hg.1 | TC1800007360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800150491.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150491.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150493.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150493.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150493.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150493.hg.1 | TC1800007360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800150493.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150493.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150493.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150490.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150490.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150490.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150490.hg.1 | TC1800007360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800150490.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150490.hg.1 | ENST00000592334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150490.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150492.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150492.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150492.hg.1 | TC1800007360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800150492.hg.1 | ENST00000262094.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800150492.hg.1 | NM_004163.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008742.hg.1 | ENST00000588466.1 | Ensembl |

RAB27B
Genomic Map