Assay Details
| Gene Symbol: FAM127A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078171.1 | NP_001071639.1 | 1 | 1133 | 96 | |
| GenBank mRNA | AF038168.1 | - | 1 | 1069 | 96 | |
| AF052096.1 | - | 1 | 1069 | 96 | ||
| BC002385.2 | - | 1 | 1063 | 2961665 | 96 | |
| BC002410.2 | - | 1 | 1063 | 2961641 | 96 | |
| CD367165.1 | - | 1 | 96 | |||
| Y13374.1 | - | 1 | 1078 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703198 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17400012 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17400009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17400014 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971292 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971272 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17400019 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971276 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971294 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703210 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971303 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17400010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971293 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17400006 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971278 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971288 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703185 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17400005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11971290 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32703213 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 43377 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P85236 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P156566 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 24419 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 7297 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P101629 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3230985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32703177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971275 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32703163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32703177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32703163 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971275 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11971273 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P156566 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P101629 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P85236 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P120968 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P156566 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P85236 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 59190 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 82438 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971293 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971289 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971304 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17399987 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17399994 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17400011 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971301 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17399999 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17400015 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971281 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971265 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971287 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971284 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971273 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971268 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971295 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971279 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971283 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17400005 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971299 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971288 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971297 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971291 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11971271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17400008 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001078171_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8170011 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8170012 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8170010 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8170009 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 201828_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11759426_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11759425_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11715218_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11715218_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11759425_at | - | - |
| Affymetrix Human Genome U219 Array | 11759426_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 201828_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 201828_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 201828_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 201828_x_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw502161 | - | - |
| Human Genome U95 Set | 33856_at | - | - |
| Human Genome U95 Set | 33856_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 201828_x_at7 | - | - |
| Human Genome U133 Set | 201828_x_at | - | - |
| Human Genome U133 Set | 201828_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041567 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041567 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1708143 | AF038168.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1708143 | CD367165.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1708143 | CR623551.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1708143 | BC106076.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1708143 | Y13374.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1708143 | AF038168.1 | human_transcriptome |

FAM127A
Genomic Map