Assay Details
| Gene Symbol: FAM13A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002806.2 | 3 - 4 | 828 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HECT and RLD domain containing E3 ubiquitin protein ligase 3 | HERC3 | -12.9kb | Sense | Chr. 4 - 88592423 - 88709303 on NCBI Build GRCh38 | |
| Downstream | family with sequence similarity 13 member A | FAM13A |
ARHGAP48, FAM13A1 |
2.6kb | Anti-sense | Chr. 4 - 88725954 - 89111398 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000487.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04007114.hg.1 | TC04000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007114.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007114.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000611121.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | ENST00000612555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007115.hg.1 | TC04000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007115.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007115.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007119.hg.1 | TC04000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007119.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007119.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007119.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007117.hg.1 | TC04000487.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007117.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007117.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04007117.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155879.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155879.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155879.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155879.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155879.hg.1 | TC0400008110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400155887.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155887.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155887.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155887.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155887.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155887.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155887.hg.1 | TC0400008110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000611121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | ENST00000612555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008110.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155883.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155883.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155883.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155883.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155883.hg.1 | TC0400008110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400155880.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155880.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155880.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155880.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155880.hg.1 | TC0400008110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400155881.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155881.hg.1 | ENST00000612555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155881.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155881.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155881.hg.1 | ENST00000612555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155881.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155881.hg.1 | TC0400008110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | ENST00000611121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | ENST00000500765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | ENST00000511543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | ENST00000611121.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | NR_002806.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400155891.hg.1 | TC0400008110.hg.1 | Clariom D TC |

FAM13A-AS1
Genomic Map