Assay Details
| Gene Symbol: LOC100129572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096041.1 | - | 1 | 1650 | 112 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein arginine methyltransferase 9 | PRMT9 | PRMT10 | -46.7kb | Sense | Chr. 4 - 147635626 - 147684230 on NCBI Build GRCh38 |
| Downstream | microRNA 4799 | MIR4799 | 49.6kb | Anti-sense | Chr. 4 - 147782595 - 147782668 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100129572
Genomic Map