Assay Details
| Gene Symbol: LBX1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096698.1 | - | 1 - 2 | 1641 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ladybird homeobox 1 | LBX1 |
HPX-6, HPX6, LBX1H, homeobox |
-5.7kb | Anti-sense | Chr. 10 - 101226976 - 101228960 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927419 | LOC101927419 | 14.0kb | Anti-sense | Chr. 10 - 101252821 - 101263550 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000547077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000547077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | ENST00000547077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000430651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000434878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000454527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000456391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000546988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000547077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000547077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | ENST00000547077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008678.hg.1 | NR_029380.1 | Refseq |

LBX1-AS1
Genomic Map