Assay Details
| Gene Symbol: CXCL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090482.1 | - | 1 | 2446 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 10 open reading frame 142 | C10orf142 | -87.2kb | Anti-sense | Chr. 10 - 44292750 - 44294649 on NCBI Build GRCh38 | |
| Downstream | TMEM72 antisense RNA 1 | TMEM72-AS1 | 424.5kb | Sense | Chr. 10 - 44811024 - 44959689 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000343575.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000374429.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395793.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000395795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000488591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | ENST00000496375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_000609.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001033886.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001178134.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_001277990.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010468.hg.1 | NM_199168.3 | Refseq |

CXCL12
Genomic Map