Assay Details
| Gene Symbol: PPP6R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097210.1 | - | 1 | 453 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DENN domain containing 6B | DENND6B |
AFI1B, FAM116B |
-52.0kb | Anti-sense | Chr. 22 - 50309032 - 50327088 on NCBI Build GRCh38 |
| Downstream | SET binding factor 1 | SBF1 |
CMT4B3, DENND7A, MTMR5 |
64.3kb | Anti-sense | Chr. 22 - 50445000 - 50475071 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PPP6R2
Genomic Map