Assay Details
| Gene Symbol: LOC283332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026948.1 | 1 - 2 | 52 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Fas apoptotic inhibitory molecule 2 | FAIM2 |
LFG, LFG2, NGP35, NMP35, TMBIM2 |
-7.2kb | Sense | Chr. 12 - 49866896 - 49904275 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927292 | LOC101927292 | 90.0 | Anti-sense | Chr. 12 - 49911953 - 49926339 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001486.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12019550.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019550.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019550.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019550.hg.1 | ENST00000552768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019550.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019550.hg.1 | TC12001486.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | ENST00000552768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019548.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019548.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019548.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019548.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019548.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019548.hg.1 | TC12001486.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | ENST00000552768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010644.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000552768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | ENST00000552768.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200181042.hg.1 | TC1200010644.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000546911.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000550636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000551922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | ENST00000552628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | NR_026948.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200181039.hg.1 | TC1200010644.hg.1 | Clariom D TC |

LOC283332
Genomic Map