Assay Details
| Gene Symbol: LOC100419773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123300.1 | - | 3 | 1053 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 316 | ZNF316 |
ENST00000305834, MZF-3 |
-14.9kb | Sense | Chr. 7 - 6637322 - 6658279 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 12 | ZNF12 |
GIOT-3, HZF11, KOX3, ZNF325 |
14.0kb | Anti-sense | Chr. 7 - 6688433 - 6706935 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100419773
Genomic Map