Assay Details
| Gene Symbol: SRSF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123171.1 | - | 1147 | 98 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BU568399.1 | - | 1 | Chr. 7 - 27964666 - 27965409 [+] on Build GRCh38 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | JAZF zinc finger 1 | JAZF1 |
TIP27, ZNF802 |
Anti-sense | Chr. 7 - 27830573 - 28180818 on NCBI Build GRCh38 | |
| Downstream | JAZF1 antisense RNA 1 | JAZF1-AS1 | 215.0kb | Sense | Chr. 7 - 28180457 - 28241377 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SRSF6
Genomic Map