Assay Details
| Gene Symbol: LTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000595.3 | NP_000586.2 | 3 - 4 | 367 | 81 | |
| NM_001159740.2 | NP_001153212.1 | 3 - 4 | 418 | 81 | ||
| XM_011514615.2 | XP_011512917.1 | 6 - 7 | 1302 | 81 | ||
| XM_011514616.2 | XP_011512918.1 | 4 - 5 | 1037 | 81 | ||
| XM_011514617.2 | XP_011512919.1 | 6 - 7 | 904 | 81 | ||
| XM_011514618.1 | XP_011512920.1 | 6 - 7 | 850 | 81 | ||
| GenBank mRNA | A06316.1 | - | 3 - 4 | 244 | 81 | |
| BC034729.1 | - | 2 - 3 | 211 | 4341621 | 81 | |
| BG685356.1 | - | 2 - 3 | 454 | 81 | ||
| BP338937.1 | - | 3 - 4 | 418 | 81 | ||
| CR542253.1 | - | 2 - 3 | 205 | 81 | ||
| CR542273.1 | - | 2 - 3 | 205 | 81 | ||
| D00102.1 | - | 3 - 4 | 267 | 81 | ||
| D12614.1 | - | 3 - 4 | 345 | 81 | ||
| DQ123824.1 | - | 1 - 2 | 61 | 81 | ||
| DQ123826.1 | - | 1 - 2 | 61 | 81 | ||
| X01393.1 | - | 3 - 4 | 284 | 81 | ||
More Information
Set Membership: |
|

LTA
Genomic Map