Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111939.1 | 1 - 2 | 57 | 79 | ||
| GenBank mRNA | BC020608.1 | - | 1 - 2 | 57 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC020608.1 transcripts.
Set Membership: |
|

DUSP10
Genomic Map