| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006914.hg.1 |
ENST00000578707.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400006914.hg.1 |
NR_049814.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400147465.hg.1 |
ENST00000578707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400147465.hg.1 |
NR_049814.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400147465.hg.1 |
ENST00000578707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400147465.hg.1 |
NR_049814.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400147465.hg.1 |
TC0400006914.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400006914.hg.1 |
ENST00000578707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400006914.hg.1 |
NR_049814.1 |
Refseq |