Assay Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024108.2 | NP_077013.1 | 1 - 2 | 148 | 102 | |
| XM_017027295.1 | XP_016882784.1 | 1 - 2 | 207 | 102 | ||
| GenBank mRNA | BC001907.1 | - | 1 - 2 | 132 | 3534111 | 102 |
| BC004450.1 | - | 1 - 2 | 133 | 2820830 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | TC1900010926.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190659.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |

TRAPPC6A
Genomic Map