Assay Details
| Gene Symbol: SSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120448.1 | 5 - 6 | 821 | 102 | ||
| GenBank mRNA | BQ216448.1 | - | 5 - 6 | 611 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ras responsive element binding protein 1 | RREB1 |
FINB, HNT, LZ321, RREB-1, Zep-1 |
-43.4kb | Anti-sense | Chr. 6 - 7107597 - 7251980 on NCBI Build GRCh38 |
| Downstream | cancer antigen 1 | CAGE1 |
CT3, CT95, CTAG3, bA69L16.7 |
27.8kb | Sense | Chr. 6 - 7326654 - 7389709 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SSR1
Genomic Map