Assay Details
| Gene Symbol: LOC100505635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125831.1 | 3 - 4 | 396 | 92 | ||
| GenBank mRNA | AL079669.1 | - | 3 - 4 | 370 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 6 | KIF6 |
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1 |
-163.3kb | Sense | Chr. 6 - 39335100 - 39725405 on NCBI Build GRCh38 |
| Downstream | molybdenum cofactor synthesis 1 | MOCS1 |
MIG11, MOCOD, MOCODA |
12.3kb | Sense | Chr. 6 - 39904258 - 39934551 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201723.hg.1 | TC0600014286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201725.hg.1 | TC0600014286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201724.hg.1 | TC0600014286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201722.hg.1 | TC0600014286.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000420293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000430595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000437947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000606829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | ENST00000607675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014286.hg.1 | NR_125831.1 | Refseq |

LOC100505635
Genomic Map