Assay Details
| Gene Symbol: KIF5B-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KIF5B-RET_K15R11_COSF1255_1.0 | - | 57 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIF5B-RET fusion| Gene Symbol: KIF5B-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_KIF5B-RET_K15R11_COSF1255_1.0 | - | 57 | 102 | ||
Set Membership: |
|