Assay Details
| Gene Symbol: LOC105377590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_133944.1 | 1 - 2 | 334 | 54 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 4 open reading frame 47 | C4orf47 | -21.6kb | Sense | Chr. 4 - 185405227 - 185449828 on NCBI Build GRCh38 | |
| Downstream | PDZ and LIM domain 3 | PDLIM3 | ALP | 28.4kb | Anti-sense | Chr. 4 - 185500660 - 185535606 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105377590
Genomic Map