Assay Details
| Gene Symbol: LOC105378137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134590.1 | 1 - 2 | 208 | 108 | ||
| GenBank mRNA | AW295767.1 | - | 1 - 2 | 108 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FERM domain containing 1 | FRMD1 | bA164L23.1 | -132.3kb | Sense | Chr. 6 - 168055745 - 168093007 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929420 | LOC101929420 | 16.4kb | Anti-sense | Chr. 6 - 168242938 - 168262578 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105378137
Genomic Map