Assay Details
| Gene Symbol: MIR4259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130464.1 | 1 | 49 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | V-set and immunoglobulin domain containing 8 | VSIG8 | -37.3kb | Sense | Chr. 1 - 159854316 - 159862657 on NCBI Build GRCh38 | |
| Downstream | transgelin 2 | TAGLN2 | HA1756 | 18.0kb | Sense | Chr. 1 - 159918107 - 159925542 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MIR4259
Genomic Map