Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095539.1 | - | 1 - 2 | 105 | 76 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000295225.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000360990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000377181.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000461336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000468529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000475491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | ENST00000486055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_001034914.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008464.hg.1 | NM_013434.4 | Refseq |

KCNIP3
Genomic Map