Assay Details
| Gene Symbol: RBM4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017018402.1 | XP_016873891.1 | 3 - 4 | 1224 | 96 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000528106.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000528106.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000534198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000534198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | ENST00000534198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000310046.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000524637.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000525754.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000528106.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000528106.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000529195.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531036.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000531969.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000534198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000534198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | ENST00000534198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_001286135.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011299.hg.1 | NM_031492.3 | Refseq |

RBM4B
Genomic Map