| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008036.hg.1 |
ENST00000609434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008036.hg.1 |
ENST00000595595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008036.hg.1 |
ENST00000609434.1 |
Ensembl |