Assay Details
| Gene Symbol: LOC100507210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135574.1 | 1 | 25 | 52 | ||
| GenBank mRNA | BM083293.1 | - | 2 | 52 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507210| Gene Symbol: LOC100507210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135574.1 | 1 | 25 | 52 | ||
| GenBank mRNA | BM083293.1 | - | 2 | 52 | ||
Set Membership: |
|