Assay Details
| Gene Symbol: LRP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK122887.1 | - | 1 | 246 | 102 | |
More Information
Set Membership: |
|
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRP8| Gene Symbol: LRP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK122887.1 | - | 1 | 246 | 102 | |
Set Membership: |
|