Assay Details
| Gene Symbol: UFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286703.1 | NP_001273632.1 | 2 - 3 | 214 | 108 | |
| GenBank mRNA | BI761948.1 | - | 2 - 3 | 97 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286703.1 transcripts.
Set Membership: |
|

UFM1
Genomic Map