Assay Details
| Gene Symbol: NOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185057.2 | NP_001171986.1 | 3 - 4 | 818 | 66 | |
| NM_001276307.1 | NP_001263236.1 | 5 - 6 | 852 | 66 | ||
| NM_001276309.1 | NP_001263238.1 | 3 - 4 | 712 | 66 | ||
| NM_001276311.1 | NP_001263240.1 | 3 - 4 | 704 | 66 | ||
| NM_001276312.1 | NP_001263241.1 | 5 - 6 | 1203 | 66 | ||
| NM_001276319.1 | NP_001263248.1 | 5 - 6 | 1203 | 66 | ||
| NM_003946.6 | NP_003937.1 | 3 - 4 | 828 | 66 | ||
| XM_005256217.2 | XP_005256274.1 | 3 - 4 | 1455 | 66 | ||
| XM_005256219.3 | XP_005256276.1 | 3 - 4 | 1341 | 66 | ||
| XM_017023843.1 | XP_016879332.1 | 3 - 4 | 1465 | 66 | ||
| GenBank mRNA | AF064599.1 | - | 3 - 4 | 669 | 66 | |
| AF064600.1 | - | 3 - 4 | 703 | 66 | ||
| AK294145.1 | - | 5 - 6 | 1168 | 66 | ||
| BC012798.2 | - | 3 - 4 | 714 | 3506011 | 66 | |
More Information
Set Membership: |
|

NOL3
Genomic Map