Assay Details
| Gene Symbol: LTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000595.3 | NP_000586.2 | 3 - 4 | 372 | 147 | |
| NM_001159740.2 | NP_001153212.1 | 3 - 4 | 423 | 147 | ||
| XM_011514615.2 | XP_011512917.1 | 6 - 7 | 1307 | 147 | ||
| XM_011514616.2 | XP_011512918.1 | 4 - 5 | 1042 | 147 | ||
| XM_011514617.2 | XP_011512919.1 | 6 - 7 | 909 | 147 | ||
| XM_011514618.1 | XP_011512920.1 | 6 - 7 | 855 | 147 | ||
| GenBank mRNA | A06316.1 | - | 3 - 4 | 249 | 147 | |
| BC034729.1 | - | 2 - 3 | 216 | 4341621 | 147 | |
| BG685356.1 | - | 2 - 3 | 459 | 147 | ||
| BP338937.1 | - | 3 - 4 | 423 | 147 | ||
| CR542253.1 | - | 2 - 3 | 210 | 147 | ||
| CR542273.1 | - | 2 - 3 | 210 | 147 | ||
| D00102.1 | - | 3 - 4 | 272 | 147 | ||
| D12614.1 | - | 3 - 4 | 350 | 147 | ||
| X01393.1 | - | 3 - 4 | 289 | 147 | ||
More Information
Set Membership: |
|

LTA
Genomic Map