Assay Details
| Gene Symbol: ABITRAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581115.2 | XP_005581172.1 | 1 - 2 | 360 | 76 | |
| GenBank mRNA | AB171878.1 | - | 79 | 76 | ||
More Information
Set Membership: |
|

ABITRAM
Genomic Map