Assay Details
| Gene Symbol: ANXA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283231.1 | NP_001270160.1 | 7 - 8 | 639 | 63 | |
| GenBank mRNA | AB169917.1 | - | 7 - 8 | 639 | 63 | |
| AB170204.1 | - | 7 - 8 | 627 | 63 | ||
More Information
Set Membership: |
|

ANXA5
Genomic Map