Assay Details
| Gene Symbol: THPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546568.2 | XP_005546625.1 | 4 - 5 | 1904 | 64 | |
| XM_005546572.2 | XP_005546629.1 | 4 - 5 | 1904 | 64 | ||
More Information
Set Membership: |
|

THPO
Genomic Map