Assay Details
| Gene Symbol: SIX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575974.2 | XP_005576031.2 | 1 - 2 | 1008 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIX3| Gene Symbol: SIX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575974.2 | XP_005576031.2 | 1 - 2 | 1008 | 99 | |
Set Membership: |
|