Assay Details
| Gene Symbol: A4GALT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457102.1 | XP_015312588.1 | 1 - 2 | 449 | 140 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015457102.1 transcripts.
Set Membership: |
|

A4GALT
Genomic Map