Assay Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441822.1 | XP_015297308.1 | 4 - 5 | 534 | 124 | |
| XM_015441823.1 | XP_015297309.1 | 4 - 5 | 611 | 124 | ||
| XM_015441824.1 | XP_015297310.1 | 3 - 4 | 811 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015441824.1 transcripts.
Set Membership: |
|

ZNF180
Genomic Map