Assay Details
| Gene Symbol: CACFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580515.2 | XP_005580572.1 | 4 - 5 | 2876 | 77 | |
| XM_005580517.2 | XP_005580574.1 | 4 - 5 | 523 | 77 | ||
More Information
Set Membership: |
|

CACFD1
Genomic Map