Assay Details
| Gene Symbol: WBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285167.1 | NP_001272096.1 | 3 - 4 | 347 | 81 | |
| XM_015437862.1 | XP_015293348.1 | 3 - 4 | 1255 | 81 | ||
| XM_015437863.1 | XP_015293349.1 | 4 - 5 | 572 | 81 | ||
| XM_015437864.1 | XP_015293350.1 | 3 - 4 | 1255 | 81 | ||
| GenBank mRNA | AB072784.1 | - | 3 - 4 | 689 | 81 | |
| AB170241.1 | - | 3 - 4 | 342 | 81 | ||
| AB171227.1 | - | 3 - 4 | 347 | 81 | ||
| AY775950.1 | - | 3 - 4 | 277 | 81 | ||
More Information
Set Membership: |
|

WBP2
Genomic Map