Assay Details
| Gene Symbol: C4H6orf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449328.1 | XP_015304814.1 | 1 - 2 | 1110 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4H6orf141| Gene Symbol: C4H6orf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449328.1 | XP_015304814.1 | 1 - 2 | 1110 | 92 | |