Assay Details
| Gene Symbol: Prg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008920.4 | NP_032946.1 | 5 - 6 | 665 | 74 | |
| GenBank mRNA | AK010901.1 | - | 5 - 6 | 666 | 74 | |
| AK156715.1 | - | 5 - 6 | 667 | 74 | ||
| BC109348.2 | - | 5 - 6 | 645 | 40055691 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36920 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131102 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124850 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21395610 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124850 | - | - |
| Whole Mouse Genome G4122A | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363400 | NM_008920.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363400 | AK010901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363400 | AK156715.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P363400 | BC109348.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110689 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101950 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124850 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01584363 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01584366 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01523483 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01584358 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01584357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21395603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01938376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01888497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01888496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01539707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01441267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01523484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01814433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01576737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01584359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01883200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21395588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01814431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01539709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01156183 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10473403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473405 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473402 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473400 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422873_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473399 | - | - |
| Mouse Genome 430 2.0 Array | 1422873_at | - | - |
| Mouse Genome 430 2.0 Array | 1422873_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19074_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19074_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200009826.mm.1 | NM_008920.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009826.mm.1 | NM_008920.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009826.mm.1 | TC0200001227.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200009827.mm.1 | NM_008920.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009827.mm.1 | NM_008920.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200009827.mm.1 | TC0200001227.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422873_at | - | - |
| Mouse Expression Set 430 | 1422873_PM_at | - | - |
| Murine Genome U74v2 Set | 92728_PM_at | - | - |
| Murine Genome U74v2 Set | 161048_at | - | - |
| Murine Genome U74v2 Set | 92728_at | - | - |
| Murine Genome U74v2 Set | 161048_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422873_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422873_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422873_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422873_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 438119 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4070176 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4070176 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004070176 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2613878 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2613878 | - | - |
| mouse-6 v1.1 (Illumina) | scl20686.6.1_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4070176 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20686.6.1_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4070176 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2613878 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2613878 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20686.6.1_14-S | - | - |

Prg2
Genomic Map