Assay Details
| Gene Symbol: Itk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281965.1 | NP_001268894.1 | 12 - 13 | 1354 | 81 | |
| NM_001281966.1 | NP_001268895.1 | 12 - 13 | 1345 | 81 | ||
| NM_001281968.1 | NP_001268897.1 | 12 - 13 | 1333 | 81 | ||
| NM_010583.3 | NP_034713.2 | 12 - 13 | 1336 | 81 | ||
| GenBank mRNA | AK037554.1 | - | 416 | 81 | ||
| AK083380.1 | - | 12 - 13 | 1308 | 81 | ||
| AK153613.1 | - | 12 - 13 | 1292 | 81 | ||
| BC128374.1 | - | 12 - 13 | 1262 | 40111687 | 81 | |
| BC128375.1 | - | 12 - 13 | 1259 | 40111688 | 81 | |
| D14042.1 | - | 12 - 13 | 1276 | 81 | ||
| L00619.1 | - | 12 - 13 | 1341 | 81 | ||
| L05631.1 | - | 12 - 13 | 1302 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itk
Genomic Map