Assay Details
| Gene Symbol: Tyk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205312.1 | NP_001192241.1 | 17 - 18 | 2797 | 95 | |
| NM_018793.2 | NP_061263.2 | 17 - 18 | 2728 | 95 | ||
| GenBank mRNA | AF173032.1 | - | 17 - 18 | 2663 | 95 | |
| AK154438.1 | - | 17 - 18 | 2755 | 95 | ||
| AK155923.1 | - | 17 - 18 | 2640 | 95 | ||
| AK159303.1 | - | 17 - 18 | 2697 | 95 | ||
| AK167726.1 | - | 17 - 18 | 2707 | 95 | ||
| AK170107.1 | - | 17 - 18 | 2667 | 95 | ||
| AK170940.1 | - | 17 - 18 | 2666 | 95 | ||
| AK171609.1 | - | 17 - 18 | 2640 | 95 | ||
| AK206664.1 | - | 2 - 3 | 258 | 95 | ||
| BC019789.1 | - | 1 - 2 | 144 | 95 | ||
| BC053021.1 | - | 11 - 12 | 2368 | 95 | ||
| BC094240.1 | - | 16 - 17 | 2624 | 30610201 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tyk2
Genomic Map