Assay Details
| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019494.1 | NP_062367.1 | 1 - 2 | 126 | 82 | |
| NR_038116.1 | 1 - 2 | 144 | 81 | |||
| GenBank mRNA | AF136449.1 | - | 1 - 2 | 126 | 82 | |
| AF178672.1 | - | 1 - 2 | 126 | 82 | ||
| AF179872.1 | - | 1 - 2 | 126 | 82 | ||
| AK020750.1 | - | 1 - 2 | 145 | 81 | ||
| AK040051.1 | - | 1 - 2 | 86 | 81 | ||
| AK050012.1 | - | 2333 | 81 | |||
| BC025903.1 | - | 1 - 2 | 114 | 5035983 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of leukocyte chemotaxis
chemotaxis
inflammatory response
immune response
G-protein coupled receptor signaling pathway
T cell chemotaxis
response to lipopolysaccharide
regulation of cell proliferation
cell chemotaxis
chemokine-mediated signaling pathway
cytokine activity
chemokine activity
CXCR3 chemokine receptor binding
positive regulation of leukocyte chemotaxis
chemotaxis
inflammatory response
immune response
G-protein coupled receptor signaling pathway
T cell chemotaxis
response to lipopolysaccharide
regulation of cell proliferation
cell chemotaxis
chemokine-mediated signaling pathway
cytokine activity
chemokine activity
CXCR3 chemokine receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2160387 | AF179872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2160387 | NM_019494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044413 | AF179872.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044413 | NM_019494.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173727 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044413 | AF179872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044413 | NM_019494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2160387 | AF179872.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2160387 | NM_019494.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P676403 | AK020750.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P676403 | AK040051.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P676403 | AK050012.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P676403 | AF179872.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P676403 | NM_019494.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23893805 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173727 | - | - |
| Whole Mouse Genome G4122A | A_52_P676403 | AK020750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676403 | AK040051.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676403 | AK050012.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676403 | AF179872.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676403 | NM_019494.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119585 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173727 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04630296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04630302 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04630294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04166047 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04630303 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04630295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23893799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04837575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04289050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04755242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04577043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04837577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04166045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04289055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04755244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04755243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04755245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04166047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23893792 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10531426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531425 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419697_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419698_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531420 | - | - |
| Mouse Genome 430 2.0 Array | 1419698_at | - | - |
| Mouse Genome 430 2.0 Array | 1419698_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419697_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419697_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56066_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56066_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500024327.mm.1 | TC0500002757.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500024327.mm.1 | NR_038116.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024327.mm.1 | NR_038116.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024326.mm.1 | TC0500002757.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500024326.mm.1 | NR_038116.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500024326.mm.1 | NR_038116.1 | Refseq |
| Mouse Expression Set 430 | 1419697_PM_at | - | - |
| Mouse Expression Set 430 | 1419698_at | - | - |
| Mouse Expression Set 430 | 1419698_PM_at | - | - |
| Mouse Expression Set 430 | 1419697_at | - | - |
| Murine Genome U74v2 Set | 140659_at | - | - |
| Murine Genome U74v2 Set | 140659_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419698_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419697_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419698_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419697_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 921243 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103610324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103610324 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1090551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247446 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247446 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218930 | - | - |
| mouse-6 v1.1 (Illumina) | scl26372.4_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103610324 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26372.4_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103610324 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247446 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247446 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2566439 | - | - |

Cxcl11
Genomic Map