Assay Details
| Gene Symbol: Supt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013676.1 | NP_038704.1 | 22 - 23 | 2176 | 72 | |
| GenBank mRNA | AK145117.1 | - | 22 - 23 | 2216 | 72 | |
| AK145465.1 | - | 14 - 15 | 1041 | 72 | ||
| AK146055.1 | - | 22 - 23 | 2204 | 72 | ||
| AK146453.1 | - | 22 - 23 | 2204 | 72 | ||
| AK146583.1 | - | 21 - 22 | 2423 | 72 | ||
| AK146650.1 | - | 22 - 23 | 2217 | 72 | ||
| AK159786.1 | - | 23 - 24 | 2301 | 72 | ||
| AK179916.1 | - | 2 - 3 | 147 | 72 | ||
| BC007132.1 | - | 22 - 23 | 2176 | 3496691 | 72 | |
| BC057449.1 | - | 22 - 23 | 2191 | 72 | ||
| BC058598.1 | - | 21 - 22 | 2151 | 6306109 | 72 | |
| BC059849.1 | - | 14 - 15 | 1759 | 5696323 | 72 | |
| U88539.1 | - | 22 - 23 | 2203 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt5
Genomic Map