Assay Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024945.1 | NP_001020116.1 | 10 - 11 | 1393 | 82 | |
| NM_023268.2 | NP_075757.1 | 10 - 11 | 1393 | 82 | ||
| GenBank mRNA | AB044284.1 | - | 10 - 11 | 1301 | 82 | |
| AK004880.1 | - | 10 - 11 | 1339 | 82 | ||
| AK083938.1 | - | 10 - 11 | 1344 | 82 | ||
| AK166839.1 | - | 10 - 11 | 1339 | 82 | ||
| AK169877.1 | - | 10 - 11 | 1346 | 82 | ||
| AK170449.1 | - | 10 - 11 | 1306 | 82 | ||
| BC034131.1 | - | 7 - 8 | 885 | 82 | ||
| BC076590.1 | - | 10 - 11 | 1347 | 30548238 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox1
Genomic Map