Assay Details
| Gene Symbol: Ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144828.1 | NP_659077.1 | 3 - 4 | 522 | 104 | |
| GenBank mRNA | AB221544.1 | - | 3 - 4 | 171 | 104 | |
| AY640620.1 | - | 3 - 4 | 200 | 104 | ||
| AY640621.1 | - | 2 - 3 | 658 | 104 | ||
| BC011122.1 | - | 3 - 4 | 432 | 4164563 | 104 | |
| BC026568.1 | - | 3 - 4 | 522 | 4982294 | 104 | |
| BC031129.1 | - | 3 - 4 | 270 | 4988284 | 104 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY640620.1,AY640621.1 transcripts.
Set Membership: |
|

Ppp1r1b
Genomic Map