Assay Details
| Gene Symbol: Il18bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010531.1 | NP_034661.1 | 4 - 5 | 376 | 74 | |
| GenBank mRNA | AB019505.1 | - | 2 - 3 | 225 | 74 | |
| AF110802.1 | - | 2 - 3 | 1603 | 74 | ||
| AF110803.1 | - | 3 - 4 | 303 | 74 | ||
| AF122907.1 | - | 4 - 5 | 376 | 74 | ||
| AK003370.1 | - | 4 - 5 | 392 | 74 | ||
| AK008452.1 | - | 3 - 4 | 307 | 74 | ||
| AK009721.1 | - | 3 - 4 | 303 | 74 | ||
| BC018332.1 | - | 3 - 4 | 286 | 4007411 | 74 | |
| CT010164.1 | - | 2 - 3 | 225 | 74 | ||
| CT010334.1 | - | 2 - 3 | 225 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il18bp
Genomic Map