Assay Details
| Gene Symbol: Tdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019551.2 | NP_062424.1 | 3 - 4 | 526 | 123 | |
| GenBank mRNA | AJ251328.1 | - | 3 - 4 | 500 | 123 | |
| AK009089.1 | - | 3 - 4 | 121 | |||
| AK135925.1 | - | 3 - 4 | 527 | 123 | ||
| AK167707.1 | - | 2 - 3 | 322 | 123 | ||
| BC132511.1 | - | 3 - 4 | 489 | 40130788 | 123 | |
| BC132513.1 | - | 3 - 4 | 489 | 40130790 | 123 | |
More Information
Set Membership: |
|

Tdp2
Genomic Map