Assay Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019575.4 | NP_062521.1 | 5 - 6 | 508 | 78 | |
| GenBank mRNA | AF224721.1 | - | 5 - 6 | 483 | 78 | |
| AF295102.1 | - | 5 - 6 | 501 | 78 | ||
| AK019112.1 | - | 5 - 6 | 511 | 78 | ||
| AK078226.1 | - | 5 - 6 | 499 | 78 | ||
| AK154094.1 | - | 5 - 6 | 499 | 78 | ||
| AK160755.1 | - | 5 - 6 | 499 | 78 | ||
| AK168554.1 | - | 5 - 6 | 494 | 78 | ||
| BC018215.1 | - | 5 - 6 | 477 | 3965223 | 78 | |
| BC054104.2 | - | 5 - 6 | 503 | 30021104 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284238 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284241 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284239 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284237 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284240 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172314 | BC018215.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172314 | BC054104.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172314 | AK154094.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172314 | AF224721.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172314 | NM_019575.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78492 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77358 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156140 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129971 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172314 | BC018215.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172314 | BC054104.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172314 | AK154094.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172314 | AF224721.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172314 | NM_019575.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27284237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07123058 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156140 | - | - |
| Whole Mouse Genome G4122A | A_51_P324583 | NM_001100606.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P324583 | AK020747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395727 | BC018215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395727 | BC054104.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395727 | AK154094.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395727 | AK160755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P395727 | AK168554.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164017 | BC018215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164017 | BC054104.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164017 | AK154094.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164017 | AK160755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164017 | AK168554.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104619 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123058 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129971 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08906170 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08793065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08734635 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08906167 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08734631 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08438032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08782738 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08782737 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08438048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284237 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284239 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284240 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284238 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27284241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27284274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08734634 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08438032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08354036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08438046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08354041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08954704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08771868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08438035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08954722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08624737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27284299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364820 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364814 | - | - |
| Mouse Genome 430 2.0 Array | 1430327_at | - | - |
| Mouse Genome 430 2.0 Array | 1421842_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450393_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450393_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430327_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457795_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457795_at | - | - |
| Mouse Genome 430 2.0 Array | 1421842_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56214_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56214_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000006567.mm.1 | NM_019575.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006567.mm.1 | NM_019575.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006567.mm.1 | TC1000000864.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421842_PM_a_at | - | - |
| Mouse Expression Set 430 | 1421842_a_at | - | - |
| Mouse Expression Set 430 | 1450393_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450393_a_at | - | - |
| Murine Genome U74v2 Set | 170279_r_at | - | - |
| Murine Genome U74v2 Set | 170279_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450393_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421842_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450393_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421842_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 675825 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6100079 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6100079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006100079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697270 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697270 | - | - |
| mouse-6 v1.1 (Illumina) | scl38678.9_100-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6100079 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38678.9_100-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6100079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2697270 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2697270 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38678.9_100-S | - | - |

Scamp4
Genomic Map