Assay Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027570.3 | NP_081846.3 | 5 - 6 | 648 | 92 | |
| GenBank mRNA | AY092768.1 | - | 5 - 6 | 653 | 92 | |
| BC039155.1 | - | 5 - 6 | 644 | 4242682 | 92 | |
| BC055443.1 | - | 5 - 6 | 648 | 4235345 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ldhd
Genomic Map