Assay Details
| Gene Symbol: Cd209a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133238.5 | NP_573501.1 | 4 - 5 | 380 | 111 | |
| GenBank mRNA | AF373408.1 | - | 4 - 5 | 327 | 111 | |
| AK054139.1 | - | 4 - 5 | 344 | 111 | ||
| AK171639.1 | - | 4 - 5 | 324 | 111 | ||
| AY049062.1 | - | 4 - 5 | 380 | 111 | ||
| BC111095.1 | - | 3 - 4 | 428 | 40040036 | 111 | |
More Information
Set Membership: |
|

Cd209a
Genomic Map