Assay Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145376.5 | NP_663351.3 | 7 - 8 | 765 | 67 | |
| GenBank mRNA | AB244717.2 | - | 7 - 8 | 765 | 67 | |
| AK046079.1 | - | 5 - 6 | 560 | 67 | ||
| AK046507.1 | - | 5 - 6 | 560 | 67 | ||
| AK079946.1 | - | 923 | 67 | |||
| AK081865.1 | - | 7 - 8 | 923 | 67 | ||
| AK132379.1 | - | 7 - 8 | 899 | 67 | ||
| AK155286.1 | - | 7 - 8 | 929 | 67 | ||
| AK169190.1 | - | 7 - 8 | 898 | 67 | ||
| AK170723.1 | - | 7 - 8 | 918 | 67 | ||
| AK171582.1 | - | 7 - 8 | 918 | 67 | ||
| BC005662.1 | - | 5 - 6 | 295 | 67 | ||
| BC066809.1 | - | 7 - 8 | 714 | 30475544 | 67 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK046079.1,AK046507.1,AK079946.1,BC005662.1 transcripts.
Set Membership: |
|

Lpcat1
Genomic Map